ContactUsLinkComponent
VarioPrep HPLC guard column cartridge (short) VP 15/32 NUCLEODUR - polar universal guard column cartridge - length: 15 mm, ID: 32 mm pack of 1 - compatible with VarioPrep HPLC column with ID 32 or 40 mm - for all polar NUCLEODUR and NUCLEOSIL phase 1 * 1 items
Catalog # MANA762330.320
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC guard column cartridge (short) VP 15/32 NUCLEODUR - polar universal guard column cartridge - length: 15 mm, ID: 32 mm pack of 1 - compatible with VarioPrep HPLC column with ID 32 or 40 mm - for all polar NUCLEODUR and NUCLEOSIL phase 1 * 1 items
Catalog # MANA762330.320
Supplier:  MACHEREY-NAGEL
Frequently Bought Together