ContactUsLinkComponent
VarioPrep HPLC column VP 250/32 NUCLEODUR C18 Pyramid, 5 µm *** with adjustable Endfitting *** length: 250 mm, ID: 32 mm pack of 1 1 * 1 items
Catalog # MANA762272.320
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC column VP 250/32 NUCLEODUR C18 Pyramid, 5 µm *** with adjustable Endfitting *** length: 250 mm, ID: 32 mm pack of 1 1 * 1 items
Catalog # MANA762272.320
Supplier:  MACHEREY-NAGEL
Frequently Bought Together