ContactUsLinkComponent
VarioPrep HPLC column VP 250/40 NUCLEODUR 100-5 C18 ec *** with adjustable Endfitting *** length: 250 mm, ID: 40 mm pack of 1 1 * 1 items
Catalog # MANA762022.400
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC column VP 250/40 NUCLEODUR 100-5 C18 ec *** with adjustable Endfitting *** length: 250 mm, ID: 40 mm pack of 1 1 * 1 items
Catalog # MANA762022.400
Supplier:  MACHEREY-NAGEL
Frequently Bought Together