ContactUsLinkComponent
The FRITSCH impact bar is ideal for very gentle and fast grinding of especially heat-sensitive materials such as powder coatings or plastics as well as for the smooth pre-crushing and fine comminution of hard-brittle to soft, fatty or samples with re 1 * 1 items
Catalog # FRIT14.4470.00
Supplier:  FRITSCH
The FRITSCH impact bar is ideal for very gentle and fast grinding of especially heat-sensitive materials such as powder coatings or plastics as well as for the smooth pre-crushing and fine comminution of hard-brittle to soft, fatty or samples with re 1 * 1 items
Catalog # FRIT14.4470.00
Supplier:  FRITSCH
Frequently Bought Together