ContactUsLinkComponent
Sieve ring 1.5 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample pr eparation according to RoHS null
Catalog # FRIT14.4447.32
Supplier:  FRITSCH
Sieve ring 1.5 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample pr eparation according to RoHS null
Catalog # FRIT14.4447.32
Supplier:  FRITSCH
Frequently Bought Together