ContactUsLinkComponent
Sieve ring 1 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample prep aration according to RoHS null
  FRIT14.4446.32
 :  FRITSCH
Sieve ring 1 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample prep aration according to RoHS null
  FRIT14.4446.32
 :  FRITSCH
Frequently Bought Together