ContactUsLinkComponent
Sieve ring 0.08 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample p reparation according to RoHS null
Catalog # FRIT14.4441.32
Supplier:  FRITSCH
Sieve ring 0.08 mm trapezoidal perforation with reinforced edges, pure titanium for heavy-metal and iron-free grinding and sample p reparation according to RoHS null
Catalog # FRIT14.4441.32
Supplier:  FRITSCH
Frequently Bought Together