ContactUsLinkComponent
With the screw-on grinding bowl lid made of agate, you can seal the agate grinding bowl with 420 ml, 250 ml and 125 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the glove box and the mill. 1 * 1 items
Catalog # FRIT50.6405.00
Supplier:  FRITSCH
With the screw-on grinding bowl lid made of agate, you can seal the agate grinding bowl with 420 ml, 250 ml and 125 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the glove box and the mill. 1 * 1 items
Catalog # FRIT50.6405.00
Supplier:  FRITSCH
Frequently Bought Together