ContactUsLinkComponent
With the screw-on grinding bowl lid made of hardmetal tungsten carbide, you can seal the hardmetal tungsten carbide grinding bowl wi th 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in th 1 * 1 items
Catalog # FRIT50.6585.00
Supplier:  FRITSCH
With the screw-on grinding bowl lid made of hardmetal tungsten carbide, you can seal the hardmetal tungsten carbide grinding bowl wi th 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in th 1 * 1 items
Catalog # FRIT50.6585.00
Supplier:  FRITSCH
Frequently Bought Together