ContactUsLinkComponent
Grinding bowl sintered corundum, with steel casing, 80 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9630.00
Supplier:  FRITSCH
Grinding bowl sintered corundum, with steel casing, 80 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9630.00
Supplier:  FRITSCH
Frequently Bought Together