Grinding bowls hardmetal tungsten carbide with steel casing, 250 ml volume for PULVERISETTE 6 premium line null
FRIT50.6790.00
: FRITSCH
Grinding bowls hardmetal tungsten carbide with steel casing, 250 ml volume for PULVERISETTE 6 premium line null
FRIT50.6790.00
: FRITSCH
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection