ContactUsLinkComponent
Grinding bowls hardmetal tungsten carbide with steel casing, 500 ml volume for PULVERISETTE 6 premium line null
Catalog # FRIT50.6580.00
Supplier:  FRITSCH
Grinding bowls hardmetal tungsten carbide with steel casing, 500 ml volume for PULVERISETTE 6 premium line null
Catalog # FRIT50.6580.00
Supplier:  FRITSCH
Frequently Bought Together