ContactUsLinkComponent
The premium model of the FRITSCH Disk Mill for efficient fine grinding of very hard, hard-abrasive, hard-brittle, medium-hard solid s and for metal-free grinding: Even safer due to the automatic locking of the collecting vessel and grinding chamber, 1 * 1 items
Catalog # FRIT13.3040.00
Supplier:  FRITSCH
The premium model of the FRITSCH Disk Mill for efficient fine grinding of very hard, hard-abrasive, hard-brittle, medium-hard solid s and for metal-free grinding: Even safer due to the automatic locking of the collecting vessel and grinding chamber, 1 * 1 items
Catalog # FRIT13.3040.00
Supplier:  FRITSCH
Frequently Bought Together