ContactUsLinkComponent
The automatically-operated hydraulic FRITSCH Pellet Press is ideal for fast and easy preparation of both: solid and highly permeable pellets of solid samples for X-ray fluorescence analysis or infrared spectroscopy. 1 * 1 items
Catalog # FRIT09.9520.00
Supplier:  FRITSCH
The automatically-operated hydraulic FRITSCH Pellet Press is ideal for fast and easy preparation of both: solid and highly permeable pellets of solid samples for X-ray fluorescence analysis or infrared spectroscopy. 1 * 1 items
Catalog # FRIT09.9520.00
Supplier:  FRITSCH
Frequently Bought Together