ContactUsLinkComponent
automatic dispersion unit, volume 50 ml, 100 Watt ultrasonic power for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTe c plus null
Catalog # FRIT22.8800.00
Supplier:  FRITSCH
automatic dispersion unit, volume 50 ml, 100 Watt ultrasonic power for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTe c plus null
Catalog # FRIT22.8800.00
Supplier:  FRITSCH
Frequently Bought Together