ContactUsLinkComponent
Sieve ring 0.2 mm trapezoidal perforation made of stainless steel for impact bar null
Catalog # FRIT14.4483.10
Supplier:  FRITSCH
Sieve ring 0.2 mm trapezoidal perforation made of stainless steel for impact bar null
Catalog # FRIT14.4483.10
Supplier:  FRITSCH
Frequently Bought Together