ContactUsLinkComponent
Sieve ring 2 mm round perforation with reinforced edges made of stainless steel null
Catalog # FRIT14.4361.00
Supplier:  FRITSCH
Sieve ring 2 mm round perforation with reinforced edges made of stainless steel null
Catalog # FRIT14.4361.00
Supplier:  FRITSCH
Frequently Bought Together