ContactUsLinkComponent
Is recommended, if a larger sample collection than 500 ml is required, because the sample exhauster with cyclone separator is deliv ered with a 500 ml glass bottle as standard. Also 1 litre and 2 litres of sample glasses are available. null
Catalog # FRIT83.3270.00
Supplier:  FRITSCH
Is recommended, if a larger sample collection than 500 ml is required, because the sample exhauster with cyclone separator is deliv ered with a 500 ml glass bottle as standard. Also 1 litre and 2 litres of sample glasses are available. null
Catalog # FRIT83.3270.00
Supplier:  FRITSCH
Frequently Bought Together