ContactUsLinkComponent
Sieve cassette 6 mm square perforation, made of chromium-free steel ST12 for Power Cutting Mill PULVERISETTE 25 null
Katalogové číslo FRIT45.7510.09
Dodavatel:  FRITSCH
Sieve cassette 6 mm square perforation, made of chromium-free steel ST12 for Power Cutting Mill PULVERISETTE 25 null
Katalogové číslo FRIT45.7510.09
Dodavatel:  FRITSCH
Frequently Bought Together