ContactUsLinkComponent
Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (R17me2(asym)K18ac)). Application: WB,ELISA,ChIP,ChIP-Seq ,Dot,IF 1 * 50 µG
Catalog # ABNOPAB31326
Supplier:  Abnova
Rabbit polyclonal antibody raised against synthetic peptide of Histone H3 (R17me2(asym)K18ac)). Application: WB,ELISA,ChIP,ChIP-Seq ,Dot,IF 1 * 50 µG
Catalog # ABNOPAB31326
Supplier:  Abnova
Frequently Bought Together