Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
DURAN® Safety Joints, Threaded Socket

DURAN® Safety Joints, Threaded Socket

Borosilicate glass 3.3, 130 mm, 19/26 NS, 24 mm, 2,5 mm
Catalog # SCOT286581909
Supplier:   DWK Life Sciences
DURAN® Safety Joints, Threaded Socket
Catalog # SCOT286581909
Supplier:  DWK Life Sciences
DURAN® Safety Joints, Threaded Socket
DURAN® Safety Joints, Threaded Socket

Borosilicate glass 3.3, 130 mm, 19/26 NS, 24 mm, 2,5 mm

Price
4 320,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents

NS joints with threaded socket, solid.

  • Made from Borosilicate 3.3 glass
  • Increased wall thickness
  • Threaded sleeve prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here, the danger of glass breakage and associated risks is thus minimised
  • Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range
  • Tight closure due to standard ground joint

In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN® Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN® Safety Joints offer a system to separate jammed cones and sockets. This minimises the risk of breakage and related threats. The DURAN® cores with collar are designed for this purpose with an additionally formed glass ring.

Specifications
  • Socket size:
    19/26 NS
  • Tube ext.Ø:
    24 mm
  • Wall thickness:
    2,5 mm
  • Material:
    Borosilicate glass 3.3
  • Seq:
    02
  • Total length:
    130 mm