10ml PrecisionCONTROL qPCR Master Mix with ROX with TAMRA control reagent (10x 1ml) 1 * 1.000 Tests
Catalog # PCONTROLRTAMRA
Dodavatel: PRIMERDESIGN
10ml PrecisionCONTROL qPCR Master Mix with ROX with TAMRA control reagent (10x 1ml) 1 * 1.000 Tests
Catalog # PCONTROLRTAMRA
Dodavatel: PRIMERDESIGN
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR® Facial Tissues
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Dodavatel: VWR Collection