ContactUsLinkComponent
10ml PrecisionULTRA OneStep qRT-PCR Master Mix with ROX at a reduced level and inert blue dye (10x 1ml) 1 * 1.000 Tests
Catalog # OSULTRABLUELR1
Supplier:  PRIMERDESIGN
10ml PrecisionULTRA OneStep qRT-PCR Master Mix with ROX at a reduced level and inert blue dye (10x 1ml) 1 * 1.000 Tests
Catalog # OSULTRABLUELR1
Supplier:  PRIMERDESIGN
Frequently Bought Together