ContactUsLinkComponent
Blotting Paper, N°320, 110x180 mm, White, smooth surface, cotton, 702 g/m², Thickness 2,47mm 1 * 50 items
Catalog # BINZ2.530.110180
Supplier:  Ahlstrom-Munksjö
Blotting Paper, N°320, 110x180 mm, White, smooth surface, cotton, 702 g/m², Thickness 2,47mm 1 * 50 items
Catalog # BINZ2.530.110180
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together