ContactUsLinkComponent
pleated filter Grade: 39 / N, 180 g / m² Diameter: 600 mm Packing unit: 50 pieces Characteristics: Fast filtering, extra thick, strong wet bursting, soft, extra wide pore, creped 1 * 50 items
Catalog # BINZ4-483-600
Supplier:  Ahlstrom-Munksjö
pleated filter Grade: 39 / N, 180 g / m² Diameter: 600 mm Packing unit: 50 pieces Characteristics: Fast filtering, extra thick, strong wet bursting, soft, extra wide pore, creped 1 * 50 items
Catalog # BINZ4-483-600
Supplier:  Ahlstrom-Munksjö