ContactUsLinkComponent
ASTM-Thermom., S12C, white backed, -20+102:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.445x6-8mm, durable pigment,government calibrated, with Calibration Certificate with 13 test points 1 * 1 items
Catalog # AMARA300210CB/CC13
Supplier:  Amarell
ASTM-Thermom., S12C, white backed, -20+102:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.445x6-8mm, durable pigment,government calibrated, with Calibration Certificate with 13 test points 1 * 1 items
Catalog # AMARA300210CB/CC13
Supplier:  Amarell
Frequently Bought Together