ContactUsLinkComponent
Precision therm., similar ISO 655, long, +190+310:0,5°C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x 9mm, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65074-GA
Supplier:  Amarell
Precision therm., similar ISO 655, long, +190+310:0,5°C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x 9mm, richter-top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN65074-GA
Supplier:  Amarell
Frequently Bought Together