ContactUsLinkComponent
Precision therm., similar ISO 655, long, -35+115:0,5°C, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65070-CB
Supplier:  Amarell
Precision therm., similar ISO 655, long, -35+115:0,5°C, enclosed scale, capillary tube specially coated inside, with non-wetting bl ue special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65070-CB
Supplier:  Amarell
Frequently Bought Together