ContactUsLinkComponent
Precision therm., similar ISO 655, long, -55+5:0,2°C, enclosed scale, cap. colourless, toluene filled, total immersion, max. 375x9mm , richter-top finish with button, suitable for government calibration 1 * 1 items
Catalog # AMARN65040-TOL
Supplier:  Amarell
Precision therm., similar ISO 655, long, -55+5:0,2°C, enclosed scale, cap. colourless, toluene filled, total immersion, max. 375x9mm , richter-top finish with button, suitable for government calibration 1 * 1 items
Catalog # AMARN65040-TOL
Supplier:  Amarell
Frequently Bought Together