ContactUsLinkComponent
Therm. similar Landsberger, +84+102:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, 160mm immersion, 480mm long, Richter-top finish, null
Catalog # AMARN63960-CB
Supplier:  Amarell
Therm. similar Landsberger, +84+102:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, 160mm immersion, 480mm long, Richter-top finish, null
Catalog # AMARN63960-CB
Supplier:  Amarell
Frequently Bought Together