ContactUsLinkComponent
Therm. similar Landsberger, -1+18:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non-we tting blue special liquid, 160mm immersion, 480mm long, Richter-top finish with button, null
Catalog # AMARN63950-CB/CC
Supplier:  Amarell
Therm. similar Landsberger, -1+18:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non-we tting blue special liquid, 160mm immersion, 480mm long, Richter-top finish with button, null
Catalog # AMARN63950-CB/CC
Supplier:  Amarell
Frequently Bought Together