ContactUsLinkComponent
Therm. similar Landsberger, +84+102:0,05°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, total immersion, c.300x9mm, Richter-top finish with button, null
  AMARN63810-CB/CC
 :  Amarell
Therm. similar Landsberger, +84+102:0,05°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, total immersion, c.300x9mm, Richter-top finish with button, null
  AMARN63810-CB/CC
 :  Amarell
Frequently Bought Together