ContactUsLinkComponent
Therm. similar Landsberger, +17+35:0,05°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63802-CB/CC
Supplier:  Amarell
Therm. similar Landsberger, +17+35:0,05°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63802-CB/CC
Supplier:  Amarell
Frequently Bought Together