ContactUsLinkComponent
Therm. similar Landsberger, -1+18:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liq uid, total immersion, c.300x9mm, richter-top finish with button, null
Catalog # AMARN63800-CB
Supplier:  Amarell
Therm. similar Landsberger, -1+18:0,05C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liq uid, total immersion, c.300x9mm, richter-top finish with button, null
Catalog # AMARN63800-CB
Supplier:  Amarell
Frequently Bought Together