ContactUsLinkComponent
Set Amarell-Special-Therm., -10+360:0,1°C, enclosed scale, 7 therm., in stuffed case, each therm. with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARN61640-SET
Supplier:  Amarell
Set Amarell-Special-Therm., -10+360:0,1°C, enclosed scale, 7 therm., in stuffed case, each therm. with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARN61640-SET
Supplier:  Amarell
Frequently Bought Together