Amarell-Special-Therm., +148+202:0,1°C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish, with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARN61626-GA
Supplier: Amarell
Frequently Bought Together