ContactUsLinkComponent
Amarell-Special-Therm., +98+152:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid , c.450x9,0-9,5mm, Richter-top finish, with ice point scale, null
Catalog # AMARN61624-CB/CC
Supplier:  Amarell
Amarell-Special-Therm., +98+152:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid , c.450x9,0-9,5mm, Richter-top finish, with ice point scale, null
Catalog # AMARN61624-CB/CC
Supplier:  Amarell
Frequently Bought Together