Anschütztherm., similar DIN, +98+152:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special l iquid, 175x5-6mm, total immersion, Richter-top finish, null
Katalogové číslo AMARN60164-CB
Supplier: Amarell
Frequently Bought Together