ContactUsLinkComponent
Anschütz-Therm., +98+152:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 340x5 -6mm, Richter-top finish, null
Catalog # AMARN60004-CB
Supplier:  Amarell
Anschütz-Therm., +98+152:0,1C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special liquid, 340x5 -6mm, Richter-top finish, null
Catalog # AMARN60004-CB
Supplier:  Amarell
Frequently Bought Together