ContactUsLinkComponent
Therm., similar Dr. Otte, similar DIN, 0+125:1C, enclosed scale, cap. lens front, colourless, red special liquid, 370x7mm, 16lll gl ass, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN59802-FL
Supplier:  Amarell
Therm., similar Dr. Otte, similar DIN, 0+125:1C, enclosed scale, cap. lens front, colourless, red special liquid, 370x7mm, 16lll gl ass, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARN59802-FL
Supplier:  Amarell
Frequently Bought Together