ContactUsLinkComponent
Therm., similar Dr. Otte, similar DIN, 0+60:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, 370x7mm, 16lll glass, Richter-top finish, null
Catalog # AMARN59800-CB
Supplier:  Amarell
Therm., similar Dr. Otte, similar DIN, 0+60:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, 370x7mm, 16lll glass, Richter-top finish, null
Catalog # AMARN59800-CB
Supplier:  Amarell
Frequently Bought Together