ContactUsLinkComponent
Breaking point therm. type Fraass, -50+30:1C, enclosed scale, cap. lens front, colourless, toluene, upper part 140x9-10mm, lower pa rt 260x5mm, immersion 240mm, Richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM257302-TOL
Supplier:  Amarell
Breaking point therm. type Fraass, -50+30:1C, enclosed scale, cap. lens front, colourless, toluene, upper part 140x9-10mm, lower pa rt 260x5mm, immersion 240mm, Richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM257302-TOL
Supplier:  Amarell
Frequently Bought Together