Breaking point therm. similar Fraass, -31+30:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, upper part 140x9-1 0mm, lower part 260x5mm, immersion 240mm, Richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM257300-FL
Supplier: Amarell
Frequently Bought Together