Softening point therm., similar DIN 12785, -2+80:0,2°C, similar ASTM S15C, solid stem, white backed, capillary tube specially coate d inside, with non-wetting blue special liquid, total immersion, 395x7mm, durable pigment, null
Catalog # AMARM255100-CB
Supplier: Amarell
Frequently Bought Together