ContactUsLinkComponent
Angle stem therm., similar DIN 16175, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 228 x20,5mm, lower part 125x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96244-FL
Supplier:  Amarell
Angle stem therm., similar DIN 16175, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 228 x20,5mm, lower part 125x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96244-FL
Supplier:  Amarell
Frequently Bought Together