ContactUsLinkComponent
Angle stem therm., similar DIN 16175, -100+30:1°C, cap. lens front, yellow backed, milk glass scale, toluene filled, upper part 228x 20,5mm, lower part 125x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96100-TOL
Supplier:  Amarell
Angle stem therm., similar DIN 16175, -100+30:1°C, cap. lens front, yellow backed, milk glass scale, toluene filled, upper part 228x 20,5mm, lower part 125x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96100-TOL
Supplier:  Amarell
Frequently Bought Together