ContactUsLinkComponent
Angle stem therm., similar DIN 16175, 0+300:2C, cap. prismatic, yellow backed, milk glass scale, eco-friendly, green liquid, biodeg radable, upper part 228x20,5mm, lower part 185x8mm, lower part bent behind at 90 angle 1 * 1 items
Catalog # AMART96400-EF
Supplier:  Amarell
Angle stem therm., similar DIN 16175, 0+300:2C, cap. prismatic, yellow backed, milk glass scale, eco-friendly, green liquid, biodeg radable, upper part 228x20,5mm, lower part 185x8mm, lower part bent behind at 90 angle 1 * 1 items
Catalog # AMART96400-EF
Supplier:  Amarell
Frequently Bought Together