ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, -200+30:2C, cap. prismatic, blue reflecting, milk glass scale, pentane filled, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART90950-PENT
Supplier:  Amarell
Therm. straight stem, similar DIN 16174, -200+30:2C, cap. prismatic, blue reflecting, milk glass scale, pentane filled, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART90950-PENT
Supplier:  Amarell
Frequently Bought Together