ContactUsLinkComponent
Drying oven therm., straight stem, -10+100:1C, cap. yellow backed, milk glass scale with metal clasp, red special liquid, upper par t c. 200x17mm, lower part 150x7,5mm, pin-top finish 1 * 1 items
Catalog # AMART90900-FL
Supplier:  Amarell
Drying oven therm., straight stem, -10+100:1C, cap. yellow backed, milk glass scale with metal clasp, red special liquid, upper par t c. 200x17mm, lower part 150x7,5mm, pin-top finish 1 * 1 items
Catalog # AMART90900-FL
Supplier:  Amarell
Frequently Bought Together