ContactUsLinkComponent
Industrial therm, straight stem, 0+360:2C, cap. lens front, yellow backed, milk glass scale, green special liquid, upper part c. 23 5x20mm, lower part 213x8mm, cork-glass-top finish 1 * 1 items
Catalog # AMART90414-EF
Supplier:  Amarell
Industrial therm, straight stem, 0+360:2C, cap. lens front, yellow backed, milk glass scale, green special liquid, upper part c. 23 5x20mm, lower part 213x8mm, cork-glass-top finish 1 * 1 items
Catalog # AMART90414-EF
Supplier:  Amarell
Frequently Bought Together